Learn More
Hach Company Molybdate Test Kit, Model MO-2
Supplier:  Hach Company 1419301
Encompass
Molybdates are gaining over chromates for use as corrosion inhibitors due to lower toxicity. The high cost of these treatments warrant careful control of their concentration. Kit reagents react with Molybdate to form a yellow color, which is measured with a color disc.
Catalog No. 50-199-4951