Learn More
Hach Company Molybdate Test Kit, Model MO-LR
Supplier:  Hach Company 2359300
Encompass
Molybdates are used to inhibit corrosion in industrial and utility water systems. The high cost of these treatments warrant careful control of their concentration. Kit reagents react with molybdate to form a blue color, which is measured with a color disc.
Catalog No. 50-199-5872