Learn More
Hach Company Nitrogen, Nitrite Color Disc Test Kit, Model NI-6
Supplier:  Hach Company 224000
Encompass
Utilizing the simple color disc method/ferrous sulfate, this kit measures Nitrogen, Nitrite as NO2- and contains 100 tests. Range (mg/L): 0-100/ 0-2000.
Catalog No. 50-199-4743