Learn More
Hach Company Nitrogen, Ammonia Test Kit, Model NI-8
Supplier: Hach Company 224100
Designed for testing ammonia in freshwater, this kit's color disc method offers low-cost, fast, reliable results in an easy-to-carry kit.