Learn More
Hach Company Dissolved Oxygen Test Kit, Model OX-2P
Supplier:  Hach Company 146900
Encompass
Low dissolved oxygen levels in surface water systems usually indicate serious pollution. Adequate amounts are crucial for fish life, but conversely, DO must be excluded from boiler feed water to prevent corrosion. This test kit utilizes drop count titration and contains 100 tests.Method / Chemistry: Drop count t
Catalog No. 50-199-4578