Learn More
Hach Company Ozone Test Kit, Model OZ-2
Supplier:  Hach Company 2064400
Encompass
This compact and low cost ozone test kit utilizes an easy to use color disc method. Simply grab a sample, add powder and compare sample color to the color wheel to arrive on an accurate measurement.
Catalog No. 50-199-5211