Learn More
Hach Company Professional Boiler Treatment/Boiler Feed and Cooling Water Test Kit, Model PBC-DT
Catalog No. 501995855 Shop All Hach Company Products
Encompass
Proper maintenance and optimization of boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Kit includes the tests offered in Model BT-DT (PN 2350600) plus testing capability for iron (0-2.0 mg/L), silica (0-30 mg/L), chlorine (0-3.4 mg/L), nitrite (0-80 mg/L), and total dissolved solids (TDS). The versatility and accuracy of is invaluable to boiler operators and chemical sales professionals. Comes with comprehensive manual.
Catalog No. 50-199-5855 Supplier Hach Company Supplier No. 2350700
Proper maintenance and optimization of boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. Kit includes the tests offered in Model BT-DT (PN 2350600) plus testing capability for iron (0-2.0 mg/L), silica (0-30 mg/L), chlorine (0-3.4 mg/L), nitrite (0-80 mg/L), and total dissolved solids (TDS). The versatility and accuracy of is invaluable to boiler operators and chemical sales professionals. Comes with comprehensive manual.