Learn More
Hach Company Pool Master Test Kit
Supplier: Hach Company 2315001
Hach's Pool Master Test Kit combines several tests into a single test kit. The test kit includes tests for alkalinity (20-400 mg/L), free and total chlorine (0-3.4 mg/L), hardness (20-400 mg/L), and pH (6.6-8.4 mg/L), plus specialized tests for total alkalinity, calcium hardness, and cyanuric acid.