Learn More
Hach Company Sulfate Test Kit, Model SF-1
Supplier:  Hach Company 225100
Encompass
Widely found in natural waters, sulfate may be at high levels in mine drainage. High concentrations of magnesium or sodium sulfate in water act as cathartics. This test kit provides economical determination of sulfate levels using the Extinction/Turbidimetric methods.
Catalog No. 50-199-4749