Learn More
Hach Company Silica Test Kit, Model SI-5
Supplier:  Hach Company 1455400
Encompass
Dissolved silica is found in almost all natural waters. Its presence is undesirable for many industrial uses because of scale formation. High-pressure boiler turbines are especially susceptible.
Catalog No. 50-199-5072