Learn More
Hach Company Stream Survey Test Kit
Supplier:  Hach Company 2712000
Encompass
The Stream Survey Kit tests ammonia (0-2.4 mg/L), nitrate (0-10 mg/L), dissolved oxygen (0.2-20 mg/L), pH (0-14), phosphate (total - 0-40 mg/L) and temperature (-10 to 110 C).
Catalog No. 50-199-6588