Learn More
Hach Company Sulfite Test Kit, Model SU-5
Supplier:  Hach Company 148002
Encompass
Utilizing drop count titration/iodometric to determine sulfite, this kit contains 100 tests.
Catalog No. 50-199-4584