Learn More
Hach Company Sulfite Test Kit, Model SU-DT
Supplier:  Hach Company 2063300
Encompass
Using the Digital Titrator, this kit contains 100 tests with a range of 10-800 mg/L.
Catalog No. 50-199-5198