Learn More
Hach Company Tannin/Lignin Test Kit
Supplier:  Hach Company 193701
Encompass
A product of decomposed plant material, tannin occurs in natural waters and is used for boiler and cooling tower water treatment, dyeing, ink manufacture, tanning, and paper sizing. Lignin, also a plant product, is found in paper pulp manufacturing waste. Tannin and lignin are analyzed together and reported as mg/L tannic acid.
Catalog No. 50-199-4645