Learn More
Test Kit Titrant for Water Determination, acc. to Karl Fischer (Refill Pack), Apura™, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Chemical Name or Material | Test Kit Titrant |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Packaging | Glass bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.