Learn More
Med Vet International McKesson 12-Drug Panel with Adulterants Drugs of Abuse Test, 16-5125A3, 100/Case
Catalog No. 503044701
Encompass_Preferred
Drugs of Abuse Test McKesson 12-Drug Panel with Adulterants AMP, BAR, BZO, COC, mAMP/MET, MDMA, MOP300, MTD, OXY, PCP, TCA, THC (OX, pH, SG) Urine Sample 25 Tests CLIA Waived All-in-one preliminary screening test cup CLIA-waved urine drug screening Rapid, visually read results in just 5 minutes Includes a dual-scale temperature strip to help monitor specimen tampering Limited urine exposure; fits a variety of drug test settings Tests for amphetamines, barbiturates, benzos, coke, meth Also tests for ecstasy, opiates, methadone, Oxy, PCP, antidepressants, pot Individually packaged in sealed foil pouch for added control No tipping, tilting or turning required
Catalog No. 50-304-4701 Supplier Med Vet International Supplier No. 1101520CS
Drugs of Abuse Test McKesson 12-Drug Panel with Adulterants AMP, BAR, BZO, COC, mAMP/MET, MDMA, MOP300, MTD, OXY, PCP, TCA, THC (OX, pH, SG) Urine Sample 25 Tests CLIA Waived All-in-one preliminary screening test cup CLIA-waved urine drug screening Rapid, visually read results in just 5 minutes Includes a dual-scale temperature strip to help monitor specimen tampering Limited urine exposure; fits a variety of drug test settings Tests for amphetamines, barbiturates, benzos, coke, meth Also tests for ecstasy, opiates, methadone, Oxy, PCP, antidepressants, pot Individually packaged in sealed foil pouch for added control No tipping, tilting or turning required