Learn More
Cole-Parmer Fluke TL81A Deluxe Electronic Test Lead Set with case
Supplier:  Cole-Parmer 2600296
Encompass
This deluxe electronic test lead set offers a high performance at an economical price. The TL81 kit consists of a quadfold nylon pouch containing two each -- one black and one red -- of the following items: One meter long test lead with a safety shrouded, right angle banana connector on one end and a stainless steel test probe on the other end, rated up to 10A. Silicone-insulated leads have superior flexibility in cold temperatures and resistance to high temperatures. It has an insulated alligator clip that is rated up to 5A. It also has an insulated probe tip extender that is rated up to 3A.
Catalog No. 50-993-299