Learn More
Cole-Parmer Fluke TL28A Test leads, 2-way clip set ( model no. TL26)
Supplier:  Cole-Parmer 2600228
Encompass
This flexible polyethylene tubing has pharmaceutical, biotech, and general lab applications with such characteristics as good chemical resistance, ultra-low extractables and heat-sealable, along with being translucent. It has a temperature range of -40 to 100 F or -40 to 37 C. The tubing meets USP Class VI requirements. It can sterilizeEthylene oxide or gamma irradiation.
Catalog No. 50-993-289