Learn More
Cole-Parmer Fluke TL71 Premium Digital Multimeter Right Angle Test Lead Set
Catalog No. 50993249 Shop All Cole-Parmer Products
Encompass
High performance at an economical price, this multimeter ismade of flexible silicone insulated leads that are heat and cold resistant. Distinctive comfort grip probes. Recommended for microvolt measurements. CAT III 1000 V, CAT IV 600 V, 10 A.
Catalog No. 50-993-249 Supplier Cole-Parmer Supplier No. 2000850
High performance at an economical price, this multimeter ismade of flexible silicone insulated leads that are heat and cold resistant. Distinctive comfort grip probes. Recommended for microvolt measurements. CAT III 1000 V, CAT IV 600 V, 10 A.