Learn More
Cole-Parmer Megger Replacement Lead Set for MIT300 Series
Supplier:  Cole-Parmer 2001036
Encompass
This replacement lead set has red and black leads that are 1.2 m long, along with four mm right angle plugs on the instrument ends, and four mm safety probes and crocodile clips on test ends. CAT III 1000 V
Catalog No. 50-993-469