Learn More
Cole-Parmer Fluke TL224 Flexible Silicone Test Lead Set
Supplier:  Cole-Parmer 2600225
Encompass
This flexible silicone test lead set offers a high performance at an economical price. The DMM test leads in red or black have safety shrouded, standard diameter banana plugs. There is a right angle connector on one end and straight connector on the other with a reinforced strain relief. The 1.5 meter silicone-insulated wire resists heat and cold. The UL listed set is CAT IV 600 V and CAT III 1000 V with a 10 A rating.
Catalog No. 50-993-288