Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Test mixture 2 for Apolar Capillary Columns (According to Grob for GC, ready-to-use), MilliporeSigma™ Supelco™

Catalog No. p-7187594 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7205 1 mL
11-101-7204 10 x 1 mL
11-101-7203 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 11-101-7205 Supplier MilliporeSigma Supelco Supplier No. 865011ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mixture of D(-)-2,3-butanediol, decane, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylcaproic acid, methyl decanoate, methyl laurate, methyl undecanoate, 1-octanol and dodecane in hexane and methylene chloride

TRUSTED_SUSTAINABILITY

Specifications

CAS 110-54-3
Flash Point -26°C - closed cup
MDL Number MFCD00148603
Type Analytical Standard
Quantity 1 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.