Learn More
Med Vet International Attest Rapid Readout Sterilization Biological Indicator Pack Steam, 1296, 25/Case
Catalog No. 502829596
Encompass_Preferred
Brown cap The self-contained pack includes biological indicators, test packs, auto-readers and log books and gives results in 3 hours
Catalog No. 50-282-9596 Supplier Med Vet International Supplier No. 310144CS
Brown cap The self-contained pack includes biological indicators, test packs, auto-readers and log books and gives results in 3 hours