Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Test Paper
SCI_ED

Catalog No. S60125 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Cobalt Chloride Test Paper
Lead Acetate Test Paper
Litmus Blue Test Strip
Litmus Neutral Test Strip
Litmus Red Test Strip
Methyl Violet Test Strip
Moisture Detection Test Strip
PTC Taste Test Paper
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S60125 Lead Acetate Test Paper
S60100 Cobalt Chloride Test Paper
S67172 Litmus Neutral Test Strip
S60130 PTC Taste Test Paper
S60124 Methyl Violet Test Strip
S67171 Litmus Red Test Strip
S60126 Moisture Detection Test Strip
S67170 Litmus Blue Test Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S60125

Use these test papers for a variety of applications.

  • Lead Acetate Test Paper detects Hydrogen Sulfide in water in concentrations as low as 5ppm
  • Use this test paper to test your drinking water, well water or groundwater

Specifications

Certifications/Compliance NFPA
Color Indicator Gray
For Use With (Application) Quickly detect Hydrogen Sulfide in water in concentrations as low as 5ppm.
Includes 100 strips
Quantity Pk. of 24 Vials; 100 Strips/Vial
Range 1 to 400 ppm
Storage Requirements Out of direct sunlight, kept dry, keep vial sealed
Test Method Dip strip into solution
Test Time 5 to 10 sec.
Type Lead Acetate Test Paper
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.