Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Test Paper
SCI_ED

Catalog No. S67172 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Cobalt Chloride Test Paper
Lead Acetate Test Paper
Litmus Neutral Test Strip
Litmus Red Test Strip
Methyl Violet Test Strip
Moisture Detection Test Strip
PTC Taste Test Paper
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S67172 Litmus Neutral Test Strip
S60100 Cobalt Chloride Test Paper
S60130 PTC Taste Test Paper
S60124 Methyl Violet Test Strip
S67171 Litmus Red Test Strip
S60126 Moisture Detection Test Strip
S60125 Lead Acetate Test Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. S67172
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these test papers for a variety of applications.

  • A foolproof pH test to determine if a solution is acidic
  • The Blue Litmus test paper does not come with a color chart as it cannot provide you with a defined pH value
  • The test strip will turn red if dipped into an acidic solution (pH <7)
  • It will remain unchanged in an alkaline solution
  • We have tested the blue litmus with multiple pH buffers
  • The blue paper is blue at pH 8
  • At pH 7, the sample is maybe just a little red
  • The real change to red seems to start with pH 6
  • 5 and lower
  • Neutral Litmus test paper tests for Neutrality of a solution, or its acidity or alkalinity
  • The test strip will turn red if dipped into an acidic solution (pH <7) or blue if dipped into an alkaline solution (pH >7)
  • Red Litmus test paper is a foolproof pH test to determine if a solution is alkaline
  • The test strip will turn blue if dipped into an alkaline solution (pH >7)
  • Color chart not included

Specifications

Certifications/Compliance NFPA
Color Indicator Blue/Red
pH Range 0 to 14
For Use With (Application) Foolproof pH test to determine if a solution is acidic or alkaline.
Includes 100 strips
Quantity Pk. of 12 Vials; 100 Strips/Vial
Storage Requirements Out of direct sunlight, kept dry, keep vial sealed
Test Method Dip strip into solution
Test Time 1 to 2 sec.
Type Litmus Neutral Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.