Learn More
Micro Essential Lab Test Paper Refill
Catalog No. 154202T Shop All Micro Essential Lab Products
Change view
Click to view available options
Quantity:
50 Case
Catalog No. Quantity
15-420-2T 50 Case
1 options
Catalog No. 15-420-2T Supplier Micro Essential Lab Supplier No. CMR 245
Add to Cart
Add to Cart
Refill for Hydrion™ Micro Chlorine Tester
  • 10-200ppm free available chlorine
Specifications
For Use With (Equipment) Hydrion™ Micro Chlorine tester
Quantity 50 Case
Range 10 to 200 ppm
Type Chlorine Refill