Learn More
Cole-Parmer Pomona Electronics 5953 Retractable/Adjustable Needle Tip Test Probe Set
Catalog No. 50993261 Shop All Cole-Parmer Products
Encompass
Save time and money with this versatile kit that features a retractable piercing needle-tip probe fully insulated to within 1/8 inch of the tip. It has an adjustable tip, ranging from 0 to 3 inches or 0 to 76 mm in length. A banana plug is designed to fit most popular DMM jacks, while its four foot silicone insulated test lead wire is good for high temperature resistance and flexibility.
Catalog No. 50-993-261 Supplier Cole-Parmer Supplier No. 2000936
Save time and money with this versatile kit that features a retractable piercing needle-tip probe fully insulated to within 1/8 inch of the tip. It has an adjustable tip, ranging from 0 to 3 inches or 0 to 76 mm in length. A banana plug is designed to fit most popular DMM jacks, while its four foot silicone insulated test lead wire is good for high temperature resistance and flexibility.