Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho Test Reagents

Catalog No. OCD6801709
Change view
Click to view available options
Quantity:
300 Tests
600 Tests
Type:
Amphetamines
Anti-Streptolysin
Apolipoprotein A1
Apolipoprotein B
Barbiturates
Benzodiazepines
Caffeine
Cannabinoids
Cocaine Metabolite
Complement C3
Complement C4
Gentamicin
Haptoglobin
High Sensitivity C-Reactive Protein
IgA
IgG
IgM
Methadone
Microalbumin
Opiates
Prealbumin
Rheumatoid Factor
Tobramycin
Transferrin
Valproic Acid
Vancomycin
alpha-1-antitrypsin
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OCD6801709 300 Tests Vancomycin
OCD6802214 300 Tests Haptoglobin
OCD6801736 300 Tests Complement C4
OCD6801737 300 Tests Apolipoprotein A1
OCD6801988 300 Tests Barbiturates
OCD6801712 300 Tests Tobramycin
OCD6801738 300 Tests Apolipoprotein B
OCD6801767 300 Tests Transferrin
OCD6801739 300 Tests High Sensitivity C-Reactive Protein
OCD6801995 300 Tests Cocaine Metabolite
OCD6801711 300 Tests Gentamicin
OCD6801732 300 Tests IgA
OCD6801710 300 Tests Valproic Acid
OCD6801997 300 Tests Opiates
OCD6801740 300 Tests Microalbumin
OCD6801994 300 Tests Cannabinoids
OCD6801713 300 Tests Caffeine
OCD6801735 300 Tests Complement C3
OCD6801728 600 Tests -
OCD6801996 300 Tests Methadone
OCD6801729 300 Tests Rheumatoid Factor
OCD6802216 300 Tests alpha-1-antitrypsin
OCD6801989 300 Tests Benzodiazepines
OCD6802218 300 Tests Anti-Streptolysin
OCD6801991 300 Tests Amphetamines
OCD6802001 300 Tests -
OCD6801733 300 Tests IgG
OCD6801734 300 Tests IgM
OCD6801741 300 Tests Prealbumin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
Catalog No. OCD6801709 Supplier QuidelOrtho Supplier No. 6801709
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Certifications/Compliance Class 7, FDA
Type Vancomycin
For Use With (Application) Therapeutic and abused drug testing, specific Proteins
Quantity 300 Tests
Form Liquid Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.