Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
QuidelOrtho Test Reagents
Catalog No. OCD6801740
Change view
Click to view available options
Quantity:
300 Tests
600 Tests
Type:
Amphetamines
Anti-Streptolysin
Apolipoprotein A1
Apolipoprotein B
Barbiturates
Benzodiazepines
Caffeine
Cannabinoids
Cocaine Metabolite
Complement C3
Complement C4
Gentamicin
Haptoglobin
High Sensitivity C-Reactive Protein
IgA
IgG
IgM
Methadone
Microalbumin
Opiates
Prealbumin
Rheumatoid Factor
Tobramycin
Transferrin
Valproic Acid
Vancomycin
alpha-1-antitrypsin
29 product options available for selection
| Catalog No. | Quantity | Type |
|---|---|---|
| OCD6801740 | 300 Tests | Microalbumin |
| OCD6802214 | 300 Tests | Haptoglobin |
| OCD6801736 | 300 Tests | Complement C4 |
| OCD6801737 | 300 Tests | Apolipoprotein A1 |
| OCD6801988 | 300 Tests | Barbiturates |
| OCD6801712 | 300 Tests | Tobramycin |
| OCD6801738 | 300 Tests | Apolipoprotein B |
| OCD6801767 | 300 Tests | Transferrin |
| OCD6801739 | 300 Tests | High Sensitivity C-Reactive Protein |
| OCD6801995 | 300 Tests | Cocaine Metabolite |
| OCD6801711 | 300 Tests | Gentamicin |
| OCD6801732 | 300 Tests | IgA |
| OCD6801710 | 300 Tests | Valproic Acid |
| OCD6801997 | 300 Tests | Opiates |
| OCD6801709 | 300 Tests | Vancomycin |
| OCD6801994 | 300 Tests | Cannabinoids |
| OCD6801713 | 300 Tests | Caffeine |
| OCD6801735 | 300 Tests | Complement C3 |
| OCD6801728 | 600 Tests | - |
| OCD6801996 | 300 Tests | Methadone |
| OCD6801729 | 300 Tests | Rheumatoid Factor |
| OCD6802216 | 300 Tests | alpha-1-antitrypsin |
| OCD6801989 | 300 Tests | Benzodiazepines |
| OCD6802218 | 300 Tests | Anti-Streptolysin |
| OCD6801991 | 300 Tests | Amphetamines |
| OCD6802001 | 300 Tests | - |
| OCD6801733 | 300 Tests | IgG |
| OCD6801734 | 300 Tests | IgM |
| OCD6801741 | 300 Tests | Prealbumin |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
Specifications
Specifications
| Type | Microalbumin |
| For Use With (Application) | Therapeutic and abused drug testing, specific Proteins |
| Quantity | 300 Tests |
| Form | Liquid Ready-to-Use |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?