Learn More
Fritsch Milling And Sizing Inc Test Sieve 200/50 mm Mesh = 0.053 mm Iso 3310-1
SDP
Catalog No. 501534152
Encompass
test sieve 200/50 mm mesh = 0.053 mm ISO 3310-1
Catalog No. 50-153-4152 Supplier Fritsch Milling And Sizing Inc Supplier No. 30.6300.03
test sieve 200/50 mm mesh = 0.053 mm ISO 3310-1