Learn More
Fritsch Milling And Sizing Inc Test Sieve 400/65 mm Mesh = 12.5 mm Iso 3310-1
SDP
Catalog No. 501534224
Encompass
test sieve 400/65 mm mesh = 12.5 mm ISO 3310-1
Catalog No. 50-153-4224 Supplier Fritsch Milling And Sizing Inc Supplier No. 34.1000.02
test sieve 400/65 mm mesh = 12.5 mm ISO 3310-1