Learn More
Fritsch Milling And Sizing Inc Test Sieve 400/65 mm Mesh = #12 Astm (1.7mm)
SDP
Catalog No. 501534254
Encompass
test sieve 400/65 mm mesh = #12 ASTM (1.7mm)
Catalog No. 50-153-4254 Supplier Fritsch Milling And Sizing Inc Supplier No. 34.2810.02
test sieve 400/65 mm mesh = #12 ASTM (1.7mm)