Learn More
Fritsch Milling And Sizing Inc Test Sieve 400/65 mm Mesh = #45 Astm (0.355 mm)
SDP
Catalog No. 501534277
Encompass
test sieve 400/65 mm mesh = #45 ASTM (0.355 mm)
Catalog No. 50-153-4277 Supplier Fritsch Milling And Sizing Inc Supplier No. 34.4110.02
test sieve 400/65 mm mesh = #45 ASTM (0.355 mm)