Learn More
Fritsch Milling And Sizing Inc Test Sieve 400/65 mm Mesh = 0.112 mm Iso 3310-1
SDP
Catalog No. 501534293
Encompass
test sieve 400/65 mm mesh = 0.112 mm ISO 3310-1
Catalog No. 50-153-4293 Supplier Fritsch Milling And Sizing Inc Supplier No. 34.5100.02
test sieve 400/65 mm mesh = 0.112 mm ISO 3310-1