Learn More
Fritsch Milling And Sizing Inc Test Sieve 400/65 mm Mesh = #230 Astm (0.063mm)
SDP
Catalog No. 501534302
Encompass
test sieve 400/65 mm mesh = #230 ASTM (0.063mm)
Catalog No. 50-153-4302 Supplier Fritsch Milling And Sizing Inc Supplier No. 34.6010.02
Add to Cart
Add to Cart
test sieve 400/65 mm mesh = #230 ASTM (0.063mm)