Learn More
Fritsch Milling And Sizing Inc Test Sieve 8/2"
SDP
Supplier:  Fritsch Milling And Sizing Inc 35.1820.03
Encompass
Test sieve 8" dia., 2" height, frame and mesh wire made of stainless steelThis sieve can also be delivered in further sizes, for example: 200 mm dia., 50 mm height, 200 mm dia., 25 mm height, 100 mm dia., 40 mm height.
Catalog No. 50-153-4315