Learn More
Fritsch Milling And Sizing Inc Test Sieve 8/2"
Supplier: Fritsch Milling And Sizing Inc 35.2020.03
Test sieve 8" dia., 2" height, frame and mesh wire made of stainless steelThis sieve can also be delivered in further sizes, for example: 200 mm dia., 50 mm height, 200 mm dia., 25 mm height, 100 mm dia., 40 mm height.