Learn More
Thermo Scientific™ Test Specific Reagents for EliA™ Tests (Phadia™ 250)
Catalog No. 83105501P
Change view
Click to view available options
Type:
EliA ANA Positive Control 250
EliA APS Positive Control 250
EliA CCP Positive Control 250
EliA Celiac Positive Control 250
EliA IgG/IgM/IgA Negative Control 250
EliA RF Positive Control 250
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
83-105-501P EliA APS Positive Control 250
83-103-301P EliA ANA Positive Control 250
83-103-501P EliA CCP Positive Control 250
83-103-601P EliA Celiac Positive Control 250
83-103-701P EliA IgG/IgM/IgA Negative Control 250
83-107-001P EliA RF Positive Control 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 83-105-501P Supplier Thermo Scientific™ Supplier No. 83105501
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement reagents support the EliA enzyme immunoassay which measures autoantibodies for the diagnosis of autoimmune diseases
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Autoimmune Disease Autoantibodies
Type EliA APS Positive Control 250
For Use With (Application) EliA™ Tests (Phadia™ 250)
Quantity 6 Vials