Learn More
Hach Company Ammonia (Nitrogen) Test Strips, 0-6.0 mg/L
Supplier:  Hach Company 2755325
Encompass
Suitable for both lab and field testing, Hach Test Strips are easy to use and easy to read. Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish. Contains 25 tests.
Catalog No. 50-199-6628