Learn More
Med Vet International Blood Glucose Test Strips OneTouch Ultra Blue 50 Strips per Pack, 022896, 1200/Case
Catalog No. 503043973
Encompass_Preferred
DoubleSure technology automatically checks each blood sample twice FastDraw capillary action draws sample in less than 1 second Provide visual confirmation of adequate blood sample
Catalog No. 50-304-3973 Supplier Med Vet International Supplier No. 850708CS
Add to Cart
Add to Cart
Request a Quote
DoubleSure technology automatically checks each blood sample twice FastDraw capillary action draws sample in less than 1 second Provide visual confirmation of adequate blood sample