Learn More
Med Vet International Assure Prism Multi Blood Glucose Test Strips, 530050, 50/Box
Supplier:  Med Vet International 942353BX
Encompass_Preferred
Precise and accurate test strip chemistry is augmented by auto-coding reducing the chance of error Assure Prism multi Test Strips should be used only with fresh capillary whole blood samples Do not reuse test strips Store test strips in a cool and dry place at a temperature of 34-86?F (1-30?C)
Catalog No. 50-304-5171
 Restricted Item