Learn More
Teco Diagnostics TEST STRIP GLU/PRO/PH/SG C/PK
Catalog No. NC0133797
Encompass_Preferred
Teco Urine Reagent Strips (URS) for Urinalysis are firm plastic strips to which several different reagent areas are affixed. Depending on the product being used, Teco Urine Reagent Strips provide tests for Glucose, Specific Gravity, pH, Protein. 1x100 Strips Bottle, 1x Teco 1-11 URS Color Chart, Package Insert
Catalog No. NC0133797 Supplier Teco Diagnostics Supplier No. URS4S100
Teco Urine Reagent Strips (URS) for Urinalysis are firm plastic strips to which several different reagent areas are affixed. Depending on the product being used, Teco Urine Reagent Strips provide tests for Glucose, Specific Gravity, pH, Protein. 1x100 Strips Bottle, 1x Teco 1-11 URS Color Chart, Package Insert