Learn More
Hach Company Chloride QuanTab Test Strips, 300-6000 mg/L
Catalog No. 501996622 Shop All Hach Company Products
Encompass
Chloride is found in nearly all natural waters and affects human taste above 250 mg/L. High levels inhibit plant growth, and many industrial processes need to limit chloride concentration. Method/Chemistry: Quan Tab Test Strips. Range: 30 mg/L. Contains 40 tests.
Catalog No. 50-199-6622 Supplier Hach Company Supplier No. 2751340
Chloride is found in nearly all natural waters and affects human taste above 250 mg/L. High levels inhibit plant growth, and many industrial processes need to limit chloride concentration. Method/Chemistry: Quan Tab Test Strips. Range: 30 mg/L. Contains 40 tests.