Learn More
CoaguSense™ Test Strip Kit
Catalog No. 23654002
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
23-654-002 50
1 options
This item is not returnable. View return policy
Catalog No. 23-654-002 Supplier CoaguSense™ Supplier No. 03P5650
This item is not returnable. View return policy
CoaguSense™ Test Strip Kit is a CLIA waived test that performs like a mini fibrometer and it not sensitive to hematocrit levels.
  • 50 patient test strips
  • 2 high and 2 low control strips
  • 1 control activation solution vial
  • 54 sample transferred tubes with plungers
  • Package insert
  • Works with both PT1 and PT2 Meters
  • 9 year safety and quality record
Specifications
Content And Storage Room Temperature
Type PT/INR
Quantity 50