Learn More
Hach Company Chloride QuanTab Test Strips, 30-600 mg/L
Supplier:  Hach Company 2744940
Encompass
Chloride is found in nearly all natural waters and affects human taste above 250 mg/L. High levels inhibit plant growth, and many industrial processes need to limit chloride concentration. Contains 40 tests.
Catalog No. 50-199-6607