Learn More
Med Vet International Truemetrix Blood Glucose Test Strips, 50 Strips per Pack, R3H01-450, 24/Case
Catalog No. 503045385
Encompass_Preferred
Samples 0.5 microliters of blood For single person use only For use with TRUE METRIX blood glucose meters Triple sense technology: detects, analyzes and corrects variables
Catalog No. 50-304-5385 Supplier Med Vet International Supplier No. 984033CS
Samples 0.5 microliters of blood For single person use only For use with TRUE METRIX blood glucose meters Triple sense technology: detects, analyzes and corrects variables