Learn More
Hach Company Total Alkalinity Test Strips, 0-240 mg/L, pk/50
Supplier:  Hach Company 2744850
Encompass
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to drinking water, food, and beverage processors; boiler operators; and aquatic biologists. Contains 50 tests.
Catalog No. 50-199-6606