Learn More
Hach Company Total Hardness Test Strips, 0-425 mg/L
Supplier:  Hach Company 2745250
Encompass
Water hardness is caused almost entirely by calcium and magnesium ions. Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers. Contains 50 tests.
Catalog No. 50-199-6610