Learn More
Siemens Healthineers Test Thrombin Reagent (BFA)
Catalog No. 23044702 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 5 mL
Catalog No. Quantity
23-044-702 10 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-702 Supplier Siemens Healthineers Supplier No. 10446598
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of thrombin time in human plasma
Test Thrombin Reagent is intended for the determination of thrombin time in citrated human plasma. The reagent is suitable for monitoring of fibrinolytic therapy, screening for disorders of fibrin formation, in suspected cases of severe fibrinogen deficiency states, and differentiating between heparin-induced prolongation of the thrombin time and disorders of fibrinogen formation.
Thrombin time is found to be prolonged not only due to disorders in fibrin polymerization, but also due to the presence of heparin. Differentiation can be achieved using Batroxobin Reagent.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Thrombin Time Testing
Quantity 10 x 5 mL