Learn More
Ace Glass, Inc. Test tube, 25mL, graduated 0-25mL in 0.5mL increments, beaded rim, 19mm OD, 170mm lg
SDP
Catalog No. 50974578
Encompass_Preferred
Test tube, 25mL, graduated 0-25mL in 0.5mL increments, beaded rim, 19mm OD, 170mm lg
Catalog No. 50-974-578 Supplier Ace Glass, Inc. Supplier No. 840825
Test tube, 25mL, graduated 0-25mL in 0.5mL increments, beaded rim, 19mm OD, 170mm lg